Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30550

Sigma-Aldrich

Decane

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH3
CAS Number:
Molecular Weight:
142.28
Beilstein/REAXYS Number:
1696981
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
assay:
≥98.0% (GC)
technique(s):
GC/GC: suitable
bp:
174 °C (lit.)
vapor pressure:
1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 16.5 °C)
3.77 mmHg ( 37.7 °C)

grade

purum

assay

≥98.0% (GC)

form

liquid

autoignition temp.

410 °F

expl. lim.

2.6 %

technique(s)

GC/GC: suitable

impurities

olefins, none detected

refractive index

n20/D 1.411 (lit.)
n20/D 1.412

bp

174 °C (lit.)

mp

−30 °C (lit.)

density

0.73 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC

InChI

1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3

InChI key

DIOQZVSQGTUSAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

114.8 °F - closed cup

flash_point_c

46.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilya V Pobelov et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 24(16), 164210-164210 (2012-04-03)
We describe a new setup for simultaneous measurements of force and current in conductive nanocontacts in a liquid environment with a high sampling rate and resolution. A lab-built current-to-voltage converter allows measurements of the current over seven orders of magnitude.
Ching-Wei Njauw et al.
Langmuir : the ACS journal of surfaces and colloids, 29(12), 3879-3888 (2013-02-28)
It has been known that the addition of bile salts to lecithin organosols induces the formation of reverse wormlike micelles and that the worms are similar to long polymer chains that entangle each other to form viscoelastic solutions. In this
Ritu Ahuja et al.
Nature chemistry, 3(2), 167-171 (2011-01-25)
Aromatic hydrocarbons are among the most important building blocks in the chemical industry. Benzene, toluene and xylenes are obtained from the high temperature thermolysis of alkanes. Higher alkylaromatics are generally derived from arene-olefin coupling, which gives branched products--that is, secondary
Sivaram Pradhan et al.
Nature chemistry, 4(2), 134-139 (2012-01-25)
The one-step transformation of C(7)-C(12) linear alkanes into more valuable oxygenates provides heterogeneous catalysis with a major challenge. In evaluating the potential of a classic mixed-metal-oxide catalyst, we demonstrate new insights into the reactivity of adsorbed oxygen species. During the
Kai Sotthewes et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3662-3667 (2013-02-21)
The energetics and dynamics of the various phases of decanethiolate self-assembled monolayers on Au(111) surfaces were studied with scanning tunneling microscopy. We have observed five different phases of the decanethiolate monolayer on Au(111): four ordered phases (β, δ, χ*, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service