Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30486

Supelco

Hexadecanoic acid tryptamide

standard for GC

Synonym(s):

N-Hexadecanoyltryptamine, N-[2-(3-Indolyl)ethyl]hexadecanamide, PAT, Palmitic acid tryptamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H42N2O
CAS Number:
Molecular Weight:
398.62
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

amide
carboxylic acid

SMILES string

CCCCCCCCCCCCCCCC(=O)NCCc1c[nH]c2ccccc12

InChI

1S/C26H42N2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-19-26(29)27-21-20-23-22-28-25-18-16-15-17-24(23)25/h15-18,22,28H,2-14,19-21H2,1H3,(H,27,29)

InChI key

YZWBXGMMZOEBAN-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongbo Liang et al.
Journal of clinical medicine, 8(3) (2019-03-14)
Cardiovascular disease (CVD) is the number one cause of non-infectious morbidity and mortality in the world. The detection, measurement, and management of high blood pressure play an essential role in the prevention and control of CVDs. However, owing to the
Martin Ulrich et al.
Appetite, 107, 303-310 (2016-08-16)
The Trait and State versions of the Food Craving Questionnaire (FCQ) have been used in numerous behavioral and physiological eating studies. However, the neurobiological signature of the FCQ has not been reported yet. In the present study, 20 healthy male
Guillaume Gillot et al.
International journal of food microbiology, 241, 141-150 (2016-10-25)
Penicillium roqueforti is used as a ripening culture for blue cheeses and largely contributes to their organoleptic quality and typical characteristics. Different types of blue cheeses are manufactured and consumed worldwide and have distinct aspects, textures, flavors and colors. These
Chang-Sei Kim et al.
IEEE transactions on bio-medical engineering, 65(11), 2384-2391 (2018-07-12)
The goal was to propose and establish the proof of concept of an ultraconvenient cuffless blood pressure monitoring approach based on the ballistocardiogram. The proposed approach monitors blood pressure by exploiting two features in the whole-body head-to-foot ballistocardiogram measured using
G Campi et al.
Nanoscale, 9(44), 17274-17283 (2017-11-02)
The mineralization process is crucial to the load-bearing characteristics of the bone extracellular matrix. In this work, we have studied the spatiotemporal dynamics of mineral deposition by human bone marrow mesenchymal stem cells differentiating toward osteoblasts promoted by the presence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service