Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30420

Sigma-Aldrich

Phosphinic acid

puriss. p.a., 49.5-50.5% (alkalimetric)

Synonym(s):

Hypophosphorous acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO2
CAS Number:
Molecular Weight:
66.00
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

puriss. p.a.

concentration

49.5-50.5% (alkalimetric)

impurities

≤0.001% heavy metals (as Pb)
≤0.02% total S (as SO4)

density

1.206 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤150 mg/kg
phosphate, phosphite (as H3PO3): ≤1%

cation traces

As: ≤2 mg/kg
Ba: ≤50 mg/kg
Ca: ≤50 mg/kg
Cd: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤10 mg/kg
K: ≤10 mg/kg
Mn: ≤5 mg/kg
Na: ≤60 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤1 mg/kg

SMILES string

O[PH2]=O

InChI

1S/H3O2P/c1-3-2/h3H2,(H,1,2)

InChI key

ACVYVLVWPXVTIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of phosphinic acids: Michael additions of phosphonous acids/esters to conjugated systems.
Thottathil JK, et al.
Tetrahedron Letters, 25(42), 4741-4744 (1984)
The invention of radical reactions. Part 32. Radical deoxygenations, dehalogenations, and deaminations with dialkyl phosphites and hypophosphorous acid as hydrogen sources.
Barton DHR, et al.
The Journal of Organic Chemistry, 58(24), 6838-6842 (1993)
William W Metcalf et al.
Annual review of biochemistry, 78, 65-94 (2009-06-06)
Natural products containing carbon-phosphorus bonds (phosphonic and phosphinic acids) have found widespread use in medicine and agriculture. Recent years have seen a renewed interest in the biochemistry and biology of these compounds with the cloning of the biosynthetic gene clusters
M Collinsová et al.
Current medicinal chemistry, 7(6), 629-647 (2000-03-07)
This review summarizes our knowledge of biochemical, biological and medical applications and properties of phosphinic acid compounds. Phosphinic acid compounds (phosphinates) are derivatives of phosphinic acid H2P(O)(OH). The major attention of this article is focused on applications of phosphinates of
Hypophosphorous acid and its salts: New reagents for radical chain deoxygenation, dehalogenation and deamination.
Barton DHR, et al.
Tetrahedron Letters, 33(39), 5709-5712 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service