Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30104

Sigma-Aldrich

Potassium fluoride

puriss. p.a., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KF
CAS Number:
Molecular Weight:
58.10
Beilstein/REAXYS Number:
3902818
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99%
form:
solid

grade

puriss. p.a.

assay

≥99%

form

solid

impurities

≤0.01% free acid (as HF)
≤0.01% free alkali (as K2CO3)
≤0.1% hexafluorosilicate (as K2SiF6)

mp

858 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[F-].[K+]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium fluoride (KF) on activated alumina is a highly reactive catalyst (basic in nature) for various organic reactions. High reactivity is due to the formation of potassium hexafluoroaluminiate, potassium hydroxide and potassium aluminate on the interaction of its fluoride ion with alumina. Celite coated with potassium fluoride has been proposed as a useful reagent for the hydrogen-bond-assisted reactions. Potassium fluoride on basic alumina in acetonitrile with ultrasound has been employed for the deprotection of tert-butyldimethylsilyl ethers of phenols.

Application

A versatile fluoride ion source used in a study of ion-specific swelling and de-swelling of ampholytic polymer gels, in the room temperature oxidation of noble metals in HF, and in the measurement of electronic polarizabilities of ions in polymers of alkali halides.
Potassium fluoride may be employed for doping the natural calcium phosphate (NP) for high yield synthesis of activated alkenes.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the actual catalytic agent in potassium fluoride on activated alumina systems.
Weinstock LM, et al.
Tetrahedron Letters, 27(33), 3845-3848 (1986)
Natural phosphate doped with potassium fluoride and modified with sodium nitrate: Efficient catalysts for the Knoevenagel condensation.
Sebti S, et al.
Tetrahedron Letters, 43(10), 1813-1815 (2002)
Selective desilylation of tert-butyldimethylsilyl ethers of phenols using potassium fluoride-alumina and ultrasound.
Schmittling EA and Sawyer JS.
Tetrahedron Letters, 32(49), 7207-7210 (1991)
Potassium fluoride on celite. A versatile reagent for C-, N-, O-, and S-alkylations.
Ando T and Yamawaki J.
Chemistry Letters (Jpn), 1, 45-46 (1979)
Lirong Wen et al.
Journal of combinatorial chemistry, 11(5), 799-805 (2009-07-28)
A series of new functionalized thiochromeno[2,3-b]pyridine derivatives have been synthesized via a sequence of Knoevenagel condensation, Michael addition, cyclization, and intramolecular nucleophilic substitution, which is first catalyzed by KF/neutral Al(2)O(3) cooperated with PEG 6000 under microwave irradiation. Experimental results indicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service