Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29984

Supelco

Eicosanoic acid tryptamide

Synonym(s):

AAT, N-Arachidoyltryptamine, N-Eicosanoyltryptamine, N-[2-(3-Indolyl)ethyl]eicosanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50N2O
CAS Number:
Molecular Weight:
454.73
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCC(=O)NCCn1ccc2ccccc12

InChI

1S/C30H50N2O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23-30(33)31-25-27-32-26-24-28-21-19-20-22-29(28)32/h19-22,24,26H,2-18,23,25,27H2,1H3,(H,31,33)

InChI key

NBCVECUGDUAJNM-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tidi Hassan et al.
Scientific reports, 7(1), 13803-13803 (2017-10-25)
Alpha-1 antitrypsin (AAT) augmentation therapy involves infusion of plasma-purified AAT to AAT deficient individuals. Whether treatment affects microRNA expression has not been investigated. This study's objectives were to evaluate the effect of AAT augmentation therapy on altered miRNA expression in
Qinghua Chen et al.
International journal of cardiology, 225, 392-401 (2016-10-23)
The study was designed to investigate if endogenous sulfur dioxide (SO2) was involved in cardiomyocyte autophagy and myocardial hypertrophy stimulated by angiotensin II (Ang II). Thirty-two C57 mice were randomly divided into control, SO2, Ang II and Ang II+SO2 groups.
Anna Beckmeyer-Borowko et al.
Respiratory research, 19(1), 156-156 (2018-08-24)
The pathophysiological role of SERPINA1 in respiratory health may be more strongly determined by the regulation of its expression than by common genetic variants. A family based study of predominantly smoking adults found methylation at two Cytosine-phosphate-Guanine sites (CpGs) in
Sabine Geiger et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 27(8), 1436-1451 (2019-05-30)
Acute graft-versus-host disease is a frequent complication associated with allogeneic hematopoietic stem cell transplantation. Patients that become refractory to initial steroid treatment have a poor prognosis. apceth-201 consists of human allogeneic mesenchymal stromal cells, engineered by lentiviral transduction to express
Leontios Pappas et al.
Nature, 516(7531), 418-422 (2014-10-09)
The neutralizing antibody response to influenza virus is dominated by antibodies that bind to the globular head of haemagglutinin, which undergoes a continuous antigenic drift, necessitating the re-formulation of influenza vaccines on an annual basis. Recently, several laboratories have described

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service