Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29245

Supelco

N1-[3,5-Bis(trifluoromethyl)benzyl]-N1-methylglycinamide hydrochloride

for GC derivatization, ≥99.0% (HPLC)

Synonym(s):

2-Amino-N-[3,5-bis(trifluoromethyl)benzyl]-N-methylacetamide hydrochloride, AMACE 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12F6N2O · HCl
CAS Number:
Molecular Weight:
350.69
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

derivatization grade ((GC derivatization))
for GC derivatization

assay

≥99.0% (HPLC)

form

solid

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

SMILES string

Cl.CN(Cc1cc(cc(c1)C(F)(F)F)C(F)(F)F)C(=O)CN

InChI

1S/C12H12F6N2O.ClH/c1-20(10(21)5-19)6-7-2-8(11(13,14)15)4-9(3-7)12(16,17)18;/h2-4H,5-6,19H2,1H3;1H

InChI key

ZBROHYVGMMPRKP-UHFFFAOYSA-N

General description

N1-[3,5-Bis(trifluoromethyl)benzyl]-N1-methylglycinamide hydrochloride (AMACE 1) is an acylation reagent and is generally used in trace organic analysis.[1]

Other Notes

Nucleophilic derivatizing reagent for the very sensitive trace-organic analysis with GC-ECD-MS[2]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Shao et al.
Journal of chromatography. A, 965(1-2), 233-237 (2002-09-19)
technique that we have termed "bypass HPLC" is introduced as a noncontaminating way to define the retention time of a trace analyte for purification purposes on an HPLC column when the amount of the analyte in real samples is too
R J Lu et al.
Analytical chemistry, 72(8), 1798-1801 (2000-04-28)
Acetamide, 2-amino-N-[[3,5-bis(trifluoromethyl)phenyl]-methyl]-N-methyl-, monohydrochloride, which we have named AMACE1, was synthesized in three steps starting from N-tritylglycine. AMACE1 was coupled via its primary amine group (pKa 8.2) under aqueous conditions to four model analytes for oxidative sugar damage to DNA: glycolate
Gang Shao et al.
Analytical chemistry, 76(11), 3049-3054 (2004-05-29)
As little as 10 pg of standard glycolic acid (glycolate) was detected in a method comprising the following sequence of steps: (1) add glycolate-2,2-d(2) as an internal standard and exchange the carboxylate oxygens in hot HCl/[(18)O]water; (2) form an amide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service