Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29235

Supelco

Multielement Anion HPCE standard solution

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

bromide, 100 ppm
chloride, 100 ppm
fluoride, 100 ppm
nitrate, 100 ppm
phosphate, 100 ppm
sulfate, 100 ppm

density

1.00 g/mL at 20 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

multi-component solution

Preparation Note

prepared with high purity salts and water

Other Notes

Standard employed in the assay of anions by capillary electrophoresis

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of anions with capillary electrophoresis and indirect UV detection.
M.M. Rhemrev-Boom
Journal of Chromatography A, 680, 675-675 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service