29222
Thio-H
Synonym(s):
2-Amino-5-(5-phenyl-2-thienyl)phenol-N,N,O-triacetic acid tricesium salt
About This Item
form
solid
fluorescence
λex 332 nm; λem 500 nm in 0.1 M Tris pH 8.0, 10 mM Ca2+
SMILES string
[Cs+].[Cs+].[Cs+].[O-]C(=O)COc1cc(ccc1N(CC([O-])=O)CC([O-])=O)-c2ccc(s2)-c3ccccc3
InChI
1S/C22H19NO7S.3Cs/c24-20(25)11-23(12-21(26)27)16-7-6-15(10-17(16)30-13-22(28)29)19-9-8-18(31-19)14-4-2-1-3-5-14;;;/h1-10H,11-13H2,(H,24,25)(H,26,27)(H,28,29);;;/q;3*+1/p-3
InChI key
GBQDXALAYDCKJB-UHFFFAOYSA-K
Features and Benefits
Compared to wellknown low-affinity Ca2+ indicators Thio-H is about three times brighter. This is based on strong molar absorption as well as high quantum yields. High fluorescent output allows low indicator concentrations to be used, which decreases the cation buffering effects of indicator. The indicator offers the interesting property of being both excitation and emission rationable, thus ratiomeric measurements are possible both in excitations as emission mode. Further spectroscopic data (0.1 M Tris, pH 8.0): low [Ca2+], Exc.max. 362 nm, Em.max. 500 nm; high [Ca2+], Exc.max. 332 nm, Em.max. 500 nm
Packaging
Other Notes
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service