Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28982

Supelco

Curcumin solution

for TLC derivatization, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v))

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

liquid

concentration

~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v))

technique(s)

thin layer chromatography (TLC): suitable

refractive index

n20/D 1.363

density

0.794 g/mL at 20 °C

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-12,24-25H,13H2,1-2H3/b7-3+,8-4+

InChI key

VFLDPWHFBUODDF-FCXRPNKRSA-N

Application

Effective TLC derivatization reagent for the colorimetric detection of various boronic acids and their derivatives

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

57.2 °F

flash_point_c

14 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple and effective colorimetric technique for the detection of boronic acids and their derivatives.
Lawrence, K., et al.
Analytical Methods : Advancing Methods and Applications, 4, 2215-2217 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service