Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28642-U

Hamilton® plunger assemblies for CTC syringes

1702CTC, for use with CTC 25μL syringe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

aluminum plunger (PTFE coated)

syringe size

25 μL

feature

PTFE tip

packaging

pkg of 10 ea

manufacturer/tradename

Hamilton 203245

volume

25 μL

compatibility

for use with CTC 25μL syringe

Looking for similar products? Visit Product Comparison Guide

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service