Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28639-U

Hamilton® 1005LTN syringe

1005LTN (C-Line), volume 5 mL, needle size 22 ga (blunt tip), needle L 51 mm (2 in.), pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

blunt tip stainless steel needle (point style 3)
glass body

needle size

22 ga (blunt tip)

syringe size

5 mL

description

gastight

feature

point style blunt tip

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 203085

needle L

51 mm (2 in.)

volume

5 mL

compatibility

for use with CTC HTS PAL Instruments (HPLC Sampling)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service