Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28618-U

Hamilton® 701N syringe

701N, volume 10 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

blunt tip stainless steel (point style 3)

needle size

22s ga (blunt tip)

feature

Microliter
Syringe, Fixed needle

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 203073

needle L

51 mm (2 in.)

volume

10 μL

compatibility

for use with CTC HTS PAL Instruments (HPLC Sampling)

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.
Microliter is a trademark of Hamilton Co.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service