Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27817

Sigma-Aldrich

10-Undecenoic acid

puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%

Synonym(s):

Undecylenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8COOH
CAS Number:
Molecular Weight:
184.28
Colour Index Number:
42650
Beilstein/REAXYS Number:
1762631
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

description

peroxide value <= 10

grade

puriss.

assay

98-100.5%

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

fats, mineral oils, complies
in water soluble matter, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)

ign. residue

≤0.1% (as SO4)

refractive index

n25/D 1.447-1.448
n20/D 1.449 (lit.)

bp

137 °C/2 mmHg (lit.)

mp

23-25 °C (lit.)

iodine value

131‑138

transition temp

solidification point ≥21 °C

solubility

water: insoluble

density

0.912 g/mL at 25 °C (lit.)

suitability

corresponds for degree of unsaturation

SMILES string

OC(=O)CCCCCCCCC=C

InChI

1S/C11H20O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2H,1,3-10H2,(H,12,13)

InChI key

FRPZMMHWLSIFAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

10-Undecenoic acid undergoes selective hydroxylation with a light-activated hybrid P450 BM3 enzyme[1]. It participates in Passerini reaction with an unsaturated isocyanide to afford dendrimers[2].

Application

10-Undecenoic acid has been used in total synthesis of clavaminol-G and 1-aminoundecan-2-ol[3].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

300.2 °F - closed cup

flash_point_c

149 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Vijai Kumar Reddy et al.
European journal of medicinal chemistry, 67, 384-389 (2013-08-06)
The first total synthesis of clavaminol-G (1) and 1-aminoundecan-2-ol (2) has been achieved from 10-undecenoic acid using epoxidation, regioselective azidolysis and in situ detosylation and reduction reactions as key steps. The methodology is extended for the synthesis of 1-aminoundecan-2-ol derivatives;
Mallory Kato et al.
Bioorganic & medicinal chemistry, 22(20), 5687-5691 (2014-06-19)
We report herein the selective hydroxylation of 10-undecenoic acid with a light-activated hybrid P450 BM3 enzyme. Under previously developed photocatalytic reaction conditions, only a monohydroxylated product is detected by gas chromatography. Hydroxylation occurs exclusively at the allylic position as confirmed
Oliver Kreye et al.
Macromolecular rapid communications, 35(3), 317-322 (2013-12-21)
The combination of the Passerini reaction and olefin cross-metathesis is shown to be a very useful approach for the divergent synthesis of dendrimers. Castor oil-derived platform chemicals, such as 10-undecenoic acid and 10-undecenal, are reacted in a Passerini reaction with
Xing-Cong Li et al.
Antimicrobial agents and chemotherapy, 52(7), 2442-2448 (2008-05-07)
Our continuing effort in antifungal natural product discovery has led to the identification of five 6-acetylenic acids with chain lengths from C(16) to C(20): 6-hexadecynoic acid (compound 1), 6-heptadecynoic acid (compound 2), 6-octadecynoic acid (compound 3), 6-nonadecynoic acid (compound 4)
Bin Guan et al.
Scientific reports, 5, 12641-12641 (2015-08-01)
This Report presents a nitrogen-doping method by chemically forming self-assembled monolayers on silicon. Van der Pauw technique, secondary-ion mass spectroscopy and low temperature Hall effect measurements are employed to characterize the nitrogen dopants. The experimental data show that the diffusion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service