Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27709

Supelco

CPC-Silica Carrier Silane Coated

30-45 mesh particle size

Synonym(s):

3-Aminopropyl-CPC-Silica carrier, Ceramic carrier, Silica carrier silane coated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

form

powder

technique(s)

LPLC: suitable

matrix

Silica

matrix active group

SAX

particle size

30-45 mesh

pore size

375 Å pore diameter

separation technique

anion exchange

General description

derivatized with 3-aminopropyl-triethoxysilane

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service