27544
Clostridial Differential Broth
suitable for microbiology, NutriSelect® Basic
Synonym(s):
DRCM, Differential Reinforced Clostridial Broth
About This Item
Recommended Products
sterility
non-sterile
Quality Level
form
powder
shelf life
limited shelf life, expiry date on the label
composition
ammonium ferric citrate, 0.5 g/L
casein peptone, 5.0 g/L
L-cysteine hydrochloride, 0.5 g/L
D(+)-glucose, 1.0 g/L
meat extract, 8.0 g/L
meat peptone, 5.0 g/L
resazurine sodium salt, 0.002 g/L
sodium acetate, 5.0 g/L
sodium disulfite, 0.5 g/L
starch, 1.0 g/L
yeast extract, 1.0 g/L
manufacturer/tradename
NutriSelect® Basic
technique(s)
microbiological culture: suitable
final pH
7.1±0.2 (25 °C)
application(s)
bioburden testing
clinical testing
environmental
food and beverages
water monitoring
microbiology
suitability
nonselective and differential for Clostridium spp.
Application
Preparation Note
Other Notes
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Clostridium perfringens can cause contamination in undercooked or improperly sterilized canned foods and water. Learn to detect, identify, and differentiate this pathogen.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service