Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27350

Sigma-Aldrich

Cinchonidine

purum, ≥98.0% (dried material, NT)

Synonym(s):

(−)-Cinchonidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39
Beilstein/REAXYS Number:
89690
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

purum

assay

≥98.0% (dried material, NT)

optical activity

[α]20/D −106±4°, c = 1% in ethanol

impurities

≤15% hydrocinchonine (HPLC)
≤5% quinine (HPLC)

ign. residue

≤0.1%

loss

≤1% loss on drying, 110 °C

mp

204-206 °C (lit.)
~205 °C

SMILES string

O[C@@H]([C@@H]1C[C@@H]2CCN1C[C@@H]2C=C)c3ccnc4ccccc34

InChI

1S/C19H22N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h2-7,9,13-14,18-19,22H,1,8,10-12H2/t13-,14-,18-,19+/m0/s1

Inchi Key

KMPWYEUPVWOPIM-KODHJQJWSA-N

Gene Information

human ... CYP2D6(1565)

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1A

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Itoh et al.
Xenobiotica; the fate of foreign compounds in biological systems, 35(6), 561-573 (2005-09-30)
In a previous paper by the authors on RS-8359, a new selective and reversible monoamine oxidase A (MAO-A) inhibitor, it was reported that the (S)-enantiomer of RS-8359 is rapidly eliminated from rats, monkeys and humans as a result of the
Chun-Yin Zhu et al.
Chemical communications (Cambridge, England), (6)(6), 738-740 (2008-05-16)
The reaction of cinchonidine (cinchonine)-derived ammonium salts with nitroolefins in the presence of Cs2CO3 to afford optically active isoxazoline N-oxides with excellent ee and high de values has been developed.
Arkajyoti Sengupta et al.
The Journal of organic chemistry, 77(23), 10525-10536 (2012-11-17)
Mechanism and the origin of enantioselectivity in the decarboxylative protonation of α-amino malonate hemiester promoted by epicinchona-thiourea hybrid organocatalyst is established by using the DFT(M06-2X/6-311+G**//ONIOM2) computational methods. The origin of stereoselectivity rendered by this hybrid bifunctional catalyst in asymmetric protonation
Erik Schmidt et al.
Langmuir : the ACS journal of surfaces and colloids, 23(15), 8087-8093 (2007-06-23)
Cinchona alkaloids are frequently used for chiral modification of supported noble metal catalysts employed in heterogeneous enantioselective hydrogenation. In order to gain molecular insight into the surface processes occurring at the metal/liquid interface, cinchonidine (CD) adsorption on vapor-deposited Rh/Al2O3 films
Erik Schmidt et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(7), 2181-2192 (2009-12-30)
The preparation of stable metal nanoparticles requires a strong interaction between the (organic) stabilizer and the metal surface that might alter the catalytic properties. This behavior has been described as "poisoning" since the stabilizer normally decreases the catalytic activity due

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service