Skip to Content
MilliporeSigma

27220

Chrysene

purum, ≥97.0%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
EC Number:
205-923-4
UNSPSC Code:
12352103
MDL number:
Beilstein/REAXYS Number:
1909297
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0%

bp

448 °C (lit.)

mp

252-254 °C (lit.)

SMILES string

c1ccc2c(c1)ccc3c4ccccc4ccc23

InChI

1S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12H

InChI key

WDECIBYCCFPHNR-UHFFFAOYSA-N

Preparation Note

λ 268 nm, log ε 5.10


Still not finding the right product?

Explore all of our products under Chrysene




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the