Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27175

Sigma-Aldrich

Chromic acid on polymer support

extent of labeling: ~3 mmol/g CrO3 loading

Synonym(s):

Amberlite IRA-900 HCrO4-form

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

extent of labeling

~3 mmol/g CrO3 loading

Other Notes

Polymer supported reagent for the clean oxidation of primary and secondary alcohols to carbonyl compounds in high yields; Reagent for the synthesis of aldehydes and ketones from allylic and benzylic halides

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Respiratory system

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Cardillo et al.
Tetrahedron Letters, 3985-3985 (1976)
G. Cainelli et al.
Journal of the American Chemical Society, 98, 6737-6737 (1976)
L.G. Wade et al.
Journal of Chemical Education, 57, 438-438 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service