Key Documents
26707
Hamilton® plunger guide
for use with Hamilton 700, 1700 Series (25-500 μL), for use with Hamilton 7000 Series (0.5 to 5 μL)
About This Item
UNSPSC Code:
41122001
material
PTFE hardware
feature
Prevents syringe plunger bending
packaging
pack of 1
manufacturer/tradename
Hamilton 14906
compatibility
for use with Hamilton 700, 1700 Series (25-500 μL)
for use with Hamilton 7000 Series (0.5 to 5 μL)
General description
Plunger guides are recommended for consistent, reproducible injections.
  • Prevents syringe plunger bending
  • Adjustable stop prevents plunger damage
  • Easily installed
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)