26290-U
Hamilton® SampleLock syringe
1100SL, volume 100 mL, needle size 22 ga (bevel tip), needle L 51 mm (2 in.)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
22 ga (bevel tip)
feature
syringe type Sample Lock
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 86346
needle L
51 mm (2 in.)
volume
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Twist valve locks the sample in the syringe
- Positive rear plunger stop eliminates sample loss and plunger blowout at high pressures (not applicable to 1705 and 1710 models)
- Connects to either male or female luer fittings by using an adapter
- Male and female luer and male luer lock adapters thread onto the SampleLock valve making the syringe compatible with many connectors and fittings
- All adapters have a Kel-F (CTFE) fluid path, with a port diameter of 0.039 in.
- Internal standards can be added through the valve port into the sample contained in the syringe
Other Notes
Plunger Assemblies are available for the 1705SL (Cat. No. 26152-U) and 1710SL (Cat. No. 26154) syringes.
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service