Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26001

Sigma-Aldrich

4-Chlorophenyl isothiocyanate

purum, ≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NCS
CAS Number:
Molecular Weight:
169.63
Beilstein/REAXYS Number:
471610
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (AT)

bp

135-136 °C/24 mmHg (lit.)

mp

42-44 °C (lit.)
43-45 °C

SMILES string

Clc1ccc(cc1)N=C=S

InChI

1S/C7H4ClNS/c8-6-1-3-7(4-2-6)9-5-10/h1-4H

InChI key

MZZVFXMTZTVUFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficacy of p-chlorophenylisothiocyanate (Sch 20350) against parasites of ruminants and horses.
E Panitz et al.
The Journal of parasitology, 67(6), 964-964 (1981-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service