Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25720

Supelco

Chloroparaffin

Selectophore, 60% chlorine basis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
26111700

product line

Selectophore

Quality Level

assay

60% chlorine basis

refractive index

n20/D 1.525

solubility

THF: 0.1 g/mL, clear

density

1.387 g/mL at 20 °C (lit.)

General description

Visit our Sensor Applications portal to learn more.

Application

Chloroparaffin may be used as a plasticizer, along with PVC during fabrication of a polymer film, used to enrich the chlorinated pesticides and concentrates them, during the determination of pesticides in water, using ATR-FTIR spectroscopy.[1] It may also be uses as a fire-retardant.[2]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Wyatt et al.
Toxicology, 77(1-2), 81-90 (1993-01-29)
Male rats and mice were administered chlorinated paraffins (CPs) by daily gavage in corn oil for 14 days. Chlorowax 500C (short chain CP with 58% chlorination), Cereclor 56L (short chain CP with 56% chlorination) and Chlorparaffin 40G (medium chain CP
Determination of pesticides in water using ATR-FTIR spectroscopy on PVC/chloroparaffin coatings.
Regan F
Analytica Chimica Acta, 334 (1), 85-92 (1996)
Mechanism of action of a fire retardant chloroparaffin.
Costa L and Camino G.
Polymer Degradation and Stability, 12 (2), 125-129 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service