About This Item
grade
ACS reagent
assay
≥99.0%
form
powder
reaction suitability
reagent type: catalyst
core: zinc
impurities
≤0.01% S compounds
≤0.01% insol. dil. H2SO4
anion traces
chloride (Cl-): ≤0.001%
nitrate (NO3-): ≤0.003%
cation traces
Ca: ≤0.005%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Mn: ≤5 ppm
Na: ≤0.05%
Pb: ≤0.005%
SMILES string
O=[Zn]
InChI
1S/O.Zn
InChI key
XLOMVQKBTHCTTD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service