Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24920

Sigma-Aldrich

Chloroiodomethane

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2I
CAS Number:
Molecular Weight:
176.38
Beilstein/REAXYS Number:
1730802
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.0% (GC)

contains

silver wool as stabilizer

refractive index

n20/D 1.58
n20/D 1.582 (lit.)

bp

108-109 °C (lit.)

density

2.422 g/mL at 25 °C (lit.)

SMILES string

ClCI

InChI

1S/CH2ClI/c2-1-3/h1H2

InChI key

PJGJQVRXEUVAFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service