Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24879

Supelco

2-Chloro-6-(3-hydroxypropylamino)-1-phenalenone

for HPLC derivatization

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14ClNO2
CAS Number:
Molecular Weight:
287.74
MDL number:
UNSPSC Code:
41115700
PubChem Substance ID:

grade

derivatization grade ((HPLC))
for HPLC derivatization

form

solid

mp

~205 °C (dec.)

suitability

in accordance for fluorescence

SMILES string

OCCCNc1ccc2C=C(Cl)C(=O)c3cccc1c23

InChI

1S/C16H14ClNO2/c17-13-9-10-5-6-14(18-7-2-8-19)11-3-1-4-12(15(10)11)16(13)20/h1,3-6,9,18-19H,2,7-8H2

InChI key

HRTMJRGKWDPJHI-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Yoon Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(51), 32433-32442 (2020-12-09)
Ferroptosis is an iron-dependent regulated necrosis mediated by lipid peroxidation. Cancer cells survive under metabolic stress conditions by altering lipid metabolism, which may alter their sensitivity to ferroptosis. However, the association between lipid metabolism and ferroptosis is not completely understood.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service