Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24540

Sigma-Aldrich

Ethyl chloroacetate

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CO2C2H5
CAS Number:
Molecular Weight:
122.55
Beilstein/REAXYS Number:
506455
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

10 mmHg ( 38 °C)
3.3 mmHg ( 20 °C)

grade

purum

assay

≥98.0% (GC)

refractive index

n20/D 1.421 (lit.)
n20/D 1.422

bp

143 °C (lit.)

mp

−26 °C (lit.)

transition temp

solidification point ~−26 °C

density

1.145 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCl

InChI

1S/C4H7ClO2/c1-2-7-4(6)3-5/h2-3H2,1H3

InChI key

VEUUMBGHMNQHGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

129.2 °F - closed cup

flash_point_c

54 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ethylester of monochloroacetic acid.
C L Braun et al.
Contact dermatitis, 16(2), 114-115 (1987-02-01)
J Q Liu et al.
The Journal of biological chemistry, 272(6), 3363-3368 (1997-02-07)
Asp10 of L-2-haloacid dehalogenase from Pseudomonas sp. YL was proposed to act as a nucleophile to attack the alpha-carbon of L-2-haloalkanoic acids to form an ester intermediate, which is hydrolyzed by nucleophilic attack of a water molecule on the carbonyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service