Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24522

Hamilton® syringes for PerkinElmer® GC Autosystem and Clarus 500

75 ASN/PE GC, volume 5 μL, needle size 26s ga (blunt tip), needle L 70 mm, needle type, LTN (fixed)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
eCl@ss:
32070500

material

blunt tip stainless steel needle (point style 3)

needle size

26s ga (blunt tip)

feature

needle type LTN (fixed)

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 88040

needle L

70 mm

volume

5 μL

compatibility

for use with PerkinElmer GC Autosystem

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.
PerkinElmer is a registered trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service