Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24375

Sigma-Aldrich

2-Chloro-1,3-dimethylimidazolidinium hexafluorophosphate

purum, ≥98.0% (T)

Synonym(s):

2-Chloro-4,5-dihydro-1,3-dimethyl-1H-imidazolium hexafluorophosphate, CIP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10ClF6N2P
CAS Number:
Molecular Weight:
278.56
Beilstein/REAXYS Number:
5369058
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

purum

assay

≥98.0% (T)

form

solid

mp

231-233 °C (lit.)

solubility

acetonitrile: 0.1 g/mL, clear

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CN1CC[N+](C)=C1Cl

InChI

1S/C5H10ClN2.F6P/c1-7-3-4-8(2)5(7)6;1-7(2,3,4,5)6/h3-4H2,1-2H3;/q+1;-1

InChI key

CNAKHAGVVMOXFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
Diazo-transfer reagents

Reagent for synthesis of:
Cannabinoid CB1 receptor agonists
Selective small-molecule melanocortin-4 receptor agonists
Imidazoles as selective cannabinoid CB2 receptor antagonists
Cyclic alpha-peptoids
Cyclic melanotropin peptide analogues selective for the human melanocortin-4 receptor

Other Notes

Peptide coupling reagent suited for hindered amino acids, the additive HOAt is recommended

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 61, 3350-3350 (1996)
K. Akaji et al.
Tetrahedron Letters, 35, 3315-3315 (1994)
Tetrahedron Letters, 36, 9341-9341 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service