Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

234036

Sigma-Aldrich

Alizarin Blue Black B

new

Synonym(s):

Mordant Black 13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16N2Na2O9S2
CAS Number:
Molecular Weight:
610.52
Colour Index Number:
63615
EC Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

form

solid

solubility

formic acid: water (2:1): 1 mg/mL, opaque

λmax

548 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[S](=O)(=O)([O-])c1ccc(cc1)Nc2c3c(c(c(c2)Nc5ccc(cc5)[S](=O)(=O)O)[O-])C(=O)c4c(cccc4)C3=O

InChI

1S/C26H18N2O9S2.2Na/c29-24-18-3-1-2-4-19(18)25(30)23-22(24)20(27-14-5-9-16(10-6-14)38(32,33)34)13-21(26(23)31)28-15-7-11-17(12-8-15)39(35,36)37;;/h1-13,27-28,31H,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2

InChI key

VBUCPERXCODTAQ-UHFFFAOYSA-L

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service