Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

227978

Sigma-Aldrich

Hydroxylamine-O-sulfonic acid

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NOSO3H
CAS Number:
Molecular Weight:
113.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

technical grade

assay

90%

form

powder

mp

210 °C (dec.) (lit.)

SMILES string

NOS(O)(=O)=O

InChI

1S/H3NO4S/c1-5-6(2,3)4/h1H2,(H,2,3,4)

InChI key

DQPBABKTKYNPMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Methods and Reactions; 671. One-Step Conversion of Alicyclic Ketones into Lactams with Hydroxylamine-O-sulfonic Acid/Formic Acid.
Olah GA and Fung AP.
Synthesis, 07, 537-538 (1979)
Hydroxylamine-O-sulfonic Acid.
Erdik E and Saczewski J.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2013)
Takuya Miyazaki et al.
Advanced healthcare materials, 9(16), e2000538-e2000538 (2020-06-26)
Messenger RNA (mRNA) shows high therapeutic potential, though effective delivery systems are still needed for boosting its application. Nanocarriers loading mRNA via polyion complexation with block catiomers into core-shell micellar structures are promising systems for enhancing mRNA delivery. Engineering the
K Sakano et al.
Mutation research, 479(1-2), 101-111 (2001-07-27)
2-Nitropropane (2-NP), a widely used industrial solvent, is carcinogenic to rats. To clarify the mechanism of carcinogenesis by 2-NP, we investigated DNA damage by 2-NP metabolites, N-isopropylhydroxylamine (IPHA) and hydroxylamine-O-sulfonic acid (HAS), using 32P-5'-end-labelled DNA fragments obtained from genes that
K Kohda et al.
Nucleic acids symposium series, (17)(17), 145-148 (1986-01-01)
Amination of guanosine (Guo) with 2,4-dinitrophenoxyamine in aqueous DMF gave 7-amino-Guo, which was readily converted to 8,5'-O-cyclo-Guo, and 8-hydroxy-Guo. Deoxyguanosine (dG) gave only deglycosylated 7-amino-G under the same reaction condition. Aminations of Guo and dG with hydroxylamine-O-sulfonic acid above pH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service