Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

227374

Sigma-Aldrich

Tetrabromo-2-sulfobenzoic acid cyclic anhydride

Synonym(s):

4,5,6,7-Tetrabromo-2,1-benzoxathiol-3-one 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7Br4O4S
CAS Number:
Molecular Weight:
499.75
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

form

solid

mp

220-222 °C (lit.)

SMILES string

Brc1c(Br)c(Br)c2c(c1Br)C(=O)OS2(=O)=O

InChI

1S/C7Br4O4S/c8-2-1-6(5(11)4(10)3(2)9)16(13,14)15-7(1)12

InChI key

QPGYGIVRJIICGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service