Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C14H13N8NaO4S3
CAS Number:
Molecular Weight:
476.49
EC Number:
248-278-4
UNSPSC Code:
51101500
MDL number:
Beilstein/REAXYS Number:
3585038
assay
≥98.0% (HPLC)
impurities
≤16% water
solubility
H2O: 50 mg/mL, clear, colorless
mode of action
cell wall synthesis | interferes
storage temp.
2-8°C
SMILES string
[Na+].Cc1nnc(SCC2=C(N3[C@H](SC2)[C@H](NC(=O)Cn4cnnn4)C3=O)C([O-])=O)s1
InChI
1S/C14H14N8O4S3.Na/c1-6-17-18-14(29-6)28-4-7-3-27-12-9(11(24)22(12)10(7)13(25)26)16-8(23)2-21-5-15-19-20-21;/h5,9,12H,2-4H2,1H3,(H,16,23)(H,25,26);/q;+1/p-1/t9-,12-;/m1./s1
InChI key
FLKYBGKDCCEQQM-WYUVZMMLSA-M
General description
Chemical structure: β-lactam
Still not finding the right product?
Explore all of our products under Cefazolin sodium salt
signalword
Danger
hcodes
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
