Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21352

Syringe for On-Column Injection, Stainless Steel Fixed Needle

volume 10 μL, 701SN, for use with Agilent/HP injectors, size 32 ga (blunt tip), needle L 12.5 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

blunt tip stainless steel needle (point style 3)

size

32 ga (blunt tip)

feature

blunt tip

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton

needle L

12.5 cm

volume

10 μL

compatibility

for use with Agilent/HP injectors

Looking for similar products? Visit Product Comparison Guide

General description

The fixed needle design of this Hamilton syringe minimizes dead volume. Stainless steel needle, 12.5 cm (for Agilent/HP injectors) or 7.5 cm (for Carlo Erba injectors) × 0.24 mm O.D. (32 ga.), 90° tip. Use with 0.32 mm I.D. or larger columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service