Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21110

Sigma-Aldrich

Calcium chloride hexahydrate

purum p.a., ≥99% (calc. based on dry substance, KT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 6H2O
CAS Number:
Molecular Weight:
219.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum p.a.

assay

≥99% (calc. based on dry substance, KT)

mp

30 °C

density

1.71 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.6H2O/h;2*1H;6*1H2/q+2;;;;;;;;/p-2

InChI key

QHFQAJHNDKBRBO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Caution

may also be liquid at temperatures >28°C

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoun A Gharaibeh et al.
Environmental science and pollution research international, 22(23), 18894-18905 (2015-07-25)
Irrigation with wastewater is a promising option to improve crop yields and to reduce pressure on freshwater sources. However, heavy metal concentrations in wastewater may cause health concerns. A greenhouse pot experiment was conducted in order to determine cadmium (Cd)
Tiago Sousa et al.
Journal of pharmaceutical sciences, 103(10), 3171-3175 (2014-08-06)
Azo-bonded prodrugs of 5-aminosalicylic acid (mesalazine)-sulfasalazine, balsalazide, and olsalazine, which are used in the treatment of ulcerative colitis, rely on colonic bacteria to cleave the azo bond and liberate the active drug in the large intestine. The aim of this
Katrien Maes et al.
Journal of chromatography. A, 1360, 217-228 (2014-08-26)
Obtaining maximal sensitivity of nano UHPLC-MS/MS methods is primordial to quantify picomolar concentrations of neuropeptides in microdialysis samples. Since aspecific adsorption of peptides to Eppendorf tubes, pipette tips and UHPLC vials is detrimental for method sensitivity, a strategy is presented
Sandeep Deshmukh et al.
Journal of chromatography. A, 1388, 1-8 (2015-03-01)
Accurate quantitation of compounds in samples of biological origin is often hampered by matrix interferences one of which occurs in GC-MS analysis from the presence of highly abundant phosphate. Consequently, high concentrations of phosphate need to be removed before sample

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service