Key Documents
20765
Hamilton® plunger guide
for use with Hamilton 75, 701,1701 Series (5-10μL only)
About This Item
UNSPSC Code:
41122001
packaging
pack of 1
manufacturer/tradename
Hamilton 14806
compatibility
for use with Hamilton 75, 701,1701 Series (5-10μL only)
General description
Plunger guides are recommended for consistent, reproducible injections.
  • Prevents syringe plunger bending
  • Adjustable stop prevents plunger damage
  • Easily installed
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)