Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20330

Supelco

4-tert-Butyl-2-methylphenol

≥97.0% (GC)

Synonym(s):

4-tert-Butyl-o-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
Beilstein/REAXYS Number:
1817502
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

Quality Level

assay

≥97.0% (GC)

form

solid

impurities

≤1.0% water

SMILES string

Cc1cc(ccc1O)C(C)(C)C

InChI

1S/C11H16O/c1-8-7-9(11(2,3)4)5-6-10(8)12/h5-7,12H,1-4H3

InChI key

SNKLPZOJLXDZCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service