Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20264

Sigma-Aldrich

(R)-2-tert-Butyl-6-methyl-1,3-dioxin-4-one

puriss., ≥99.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$88.40

$88.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$88.40

About This Item

Empirical Formula (Hill Notation):
C9H14O3
CAS Number:
Molecular Weight:
170.21
Beilstein/REAXYS Number:
4176869
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

$88.40


Check Cart for Availability

Request a Bulk Order

grade

puriss.

Quality Level

assay

≥99.0% (sum of enantiomers, HPLC)

form

solid

optical activity

[α]20/D −221±3°, c = 1% in chloroform

mp

60-62 °C

SMILES string

CC1=CC(=O)O[C@@H](O1)C(C)(C)C

InChI

1S/C9H14O3/c1-6-5-7(10)12-8(11-6)9(2,3)4/h5,8H,1-4H3/t8-/m1/s1

InChI key

HWPXDBBVCANLFT-MRVPVSSYSA-N

General description

"Reagent of the Year 1987"

Other Notes

Chiral building block; this derivative of acetoacetate undergoes various stereoselective reactions with nucleophiles and electrophiles[1][2][3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemische Berichte, 124, 1845-1845 (1991)
Synthesis, 484-484 (1989)
D. Seebach et al.
Journal of the American Chemical Society, 110, 4763-4763 (1988)
D. Seebach et al.
Helvetica Chimica Acta, 69, 1147-1147 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service