Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

19955

Supelco

Ditridecyl phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1,2-ditridecyl ester, Di-n-tridecyl phthalate, Phthalic acid ditridecyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H58O4
CAS Number:
Molecular Weight:
530.82
Beilstein/REAXYS Number:
2023076
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

InChI

1S/C34H58O4/c1-3-5-7-9-11-13-15-17-19-21-25-29-37-33(35)31-27-23-24-28-32(31)34(36)38-30-26-22-20-18-16-14-12-10-8-6-4-2/h23-24,27-28H,3-22,25-26,29-30H2,1-2H3

InChI key

YCZJVRCZIPDYHH-UHFFFAOYSA-N

General description

Ditridecyl phthalate is a high molecular weight compound belonging to the class of phthalic acid esters (PAEs), which are widely used in the manufacture of plastics, coatings, and cosmetics industries.[1] Ditridecyl phthalate is used in the manufacture of wires and cables.[2]

application

Ditridecyl phthalate may be used as an analytical standard in the following:
  • Investigating its effect on the activity of estrogen receptors including ERα, ERβ and the androgen receptor AR.[3]
  • Evaluating its developmental toxicity potential in prenatal Sprague-Dawley rats.[2]

It may be used as an analytical reference standard for the quantification of the analyte in food matrices using gas chromatography coupled to mass spectrometry (GC-MS).[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of di-n-butyl phthalate and other organic pollutants in Chongqing women undergoing parturition
Chen A-J, et al.
Environmental Pollution (Barking, Essex : 1987), 156(3), 849-853 (2008)
Extraction and GC-MS analysis of phthalate esters in food matrices: a review
RussoVM, et al.
Royal Society of Chemistry Advances, 5(46), 37023-37043 (2015)
Agonistic and antagonistic effects of phthalates and their urinary metabolites on the steroid hormone receptors ER?, ER?, and AR
Engel A, et al.
Toxicology Letters, 277, 54-63 (2017)
Prenatal developmental toxicity studies on diundecyl and ditridecyl phthalates in Sprague-Dawley rats
Saillenfait M-A, et al.
Reproductive Toxicology, 37, 49-55 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service