Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19915

Sigma-Aldrich

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide

≥97.0% (GC)

Synonym(s):

MTBSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CON(CH3)Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
241.33
Beilstein/REAXYS Number:
3606546
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

assay

≥97.0% (GC)

refractive index

n20/D 1.402 (lit.)

bp

172-175 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C

InChI

1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3

InChI key

QRKUHYFDBWGLHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Powerful reagent for the introduction of the tert-butyldimethylsilyl group under neutral conditions [1][2][3][4]

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

113.0 °F - closed cup

flash_point_c

45 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
A.C. Bazan, D.R. Knapp
Journal of Chromatography A, 236, 201-201 (1982)
M. Donike et al.
Journal of Chromatography A, 202, 483-483 (1980)
K. Schoene et al.
Fresenius Journal of Analytical Chemistry, 348, 364-364 (1994)
Xiaowei Wang et al.
Journal of chromatography. A, 1216(35), 6267-6273 (2009-07-31)
A sample pretreatment method for the determination of 18 chlorophenols (CPs) in aqueous samples by derivatization liquid-phase microextraction (LPME) was investigated using gas chromatography-mass spectrometry. Derivatization reagent was spiked into the extraction solvent to combine derivatization and extraction into one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service