Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19692

Supelco

2-Hydroxyestradiol

analytical standard

Synonym(s):

1,3,5(10)-Estratriene-2,3-17β-triol, Estra-1,3,5(10)-triene-2,3,17-triol, NSC 61711

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O3
CAS Number:
Molecular Weight:
288.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)c(O)cc34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-11-12(14(18)4-5-17(18)21)3-2-10-8-15(19)16(20)9-13(10)11/h8-9,11-12,14,17,19-21H,2-7H2,1H3/t11-,12+,14-,17-,18-/m0/s1

InChI key

DILDHNKDVHLEQB-XSSYPUMDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS identity conforms

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rashel Mizrahi et al.
Biochimica et biophysica acta, 1840(12), 3404-3412 (2014-09-16)
Mitochondria are the major source of ATP to power sperm motility. Phosphorylation of mitochondrial proteins has been proposed as a major regulatory mechanism for mitochondrial bioenergetics. Sperm motility was measured by a computer-assisted analyzer, protein detection by western blotting, membrane
Barbara J Fuhrman et al.
The Journal of clinical endocrinology and metabolism, 99(12), 4632-4640 (2014-09-12)
The gut microbiota may influence the risk of breast cancer through effects on endogenous estrogens. The objective of the study was to investigate whether urinary estrogens and estrogen metabolites are associated with the diversity and composition of the fecal microbiome.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service