Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19620

Supelco

sec-Butylbenzene

analytical standard

Synonym(s):

2-Phenylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)CH2CH3
CAS Number:
Molecular Weight:
134.22
Beilstein/REAXYS Number:
1903902
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

4 mmHg ( 37.7 °C)

assay

≥99.5% (GC)

autoignition temp.

784 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.489 (lit.)
n20/D 1.489

bp

173-174 °C (lit.)

mp

−75.5 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCC(C)c1ccccc1

InChI

1S/C10H14/c1-3-9(2)10-7-5-4-6-8-10/h4-9H,3H2,1-2H3

InChI key

ZJMWRROPUADPEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

125.6 °F - closed cup

flash_point_c

52.0 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Kaczorek et al.
Chemosphere, 90(2), 471-478 (2012-08-29)
Pseudomonas stutzeri strain 9 was isolated from petroleum-contaminated soil. The main purpose of this study was to investigate how the long-term contact of this strain with diesel oil influences its surface and biodegradation properties. The experiments showed that the tested
Metabolites of three structural isomers of butylbenzene in the bile of rainbow trout.
J Hellou et al.
Bulletin of environmental contamination and toxicology, 44(3), 487-493 (1990-03-01)
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service