Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19490

Sigma-Aldrich

Butylamine

purum, ≥98.0% (GC)

Synonym(s):

1-Aminobutane, n-Butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NH2
CAS Number:
Molecular Weight:
73.14
Beilstein/REAXYS Number:
605269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.5 (vs air)

vapor pressure

68 mmHg ( 20 °C)

grade

purum

assay

≥98.0% (GC)

autoignition temp.

594 °F

expl. lim.

9.8 %

refractive index

n20/D 1.401 (lit.)
n20/D 1.401

bp

78 °C (lit.)

mp

−49 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

CCCCN

InChI

1S/C4H11N/c1-2-3-4-5/h2-5H2,1H3

InChI key

HQABUPZFAYXKJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

19.4 °F - closed cup

flash_point_c

-7 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Ya Wu et al.
Advanced materials (Deerfield Beach, Fla.), 31(4), e1806164-e1806164 (2018-12-01)
Liquid-based thermochromics can be incorporated into an arbitrarily shaped container and provide a visual map of the temperature changes within its volume. However, photochemical degradation, narrow temperature range of operation, and the need for stringent encapsulation processes are challenges that
Enhanced photovoltaic performance in nanoimprinted pentacene-PbS nanocrystal hybrid device.
Dissanayake DMNM, et al.
Applied Physics Letters, 92(9), Enhanced photovoltai-Enhanced photovoltai (2008)
Highly efficient near-infrared hybrid organic-inorganic nanocrystal electroluminescence device.
Bourdakos KN, et al.
Applied Physics Letters, 92(15) (2008)
Javad Gholami et al.
Luminescence : the journal of biological and chemical luminescence, 31(1), 229-235 (2015-05-29)
An N-butylamine functionalized graphene oxide nanolayer was synthesized and characterized by ultraviolet (UV)-visible spectrometry, Fourier transform infrared spectroscopy, X-ray photoelectron spectroscopy, and transmission electron microscopy. Detection of iron(III) based on photoluminescence spectroscopy was investigated. The N-butylamine functionalized graphene oxide was
Photoluminescence of CdSe Nanoparticles in the Presence of a Hole Acceptor:
n-Butylamine
Landes C, et al.
The Journal of Physical Chemistry B, 105(15), 2981-2986 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service