Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19443

Supelco

Andrograpanin

analytical standard

Synonym(s):

3,14-Dideoxyandrographolide, 3-[2-[(1R,4aS,5R,8aS)-Decahydro-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-2(5H)-furanone, ent-19-Hydroxy-8(17),13-labdadien-16,15-olide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O3
CAS Number:
Molecular Weight:
318.45
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@]1(CO)CCC[C@@]2(C)[C@H](CCC3=CCOC3=O)C(=C)CC[C@H]12

InChI

1S/C20H30O3/c1-14-5-8-17-19(2,13-21)10-4-11-20(17,3)16(14)7-6-15-9-12-23-18(15)22/h9,16-17,21H,1,4-8,10-13H2,2-3H3/t16-,17-,19+,20+/m1/s1

InChI key

WKKBRRFSRMDTJB-JYBIWHBTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Andrograpanin, diterpene lactone is a hydrolysate drug used as anti-inflammatory agent. It is a minor compound of Andrographis paniculata. It has the potential to modulate the chemokine pathway and is used in anti-infectious function.[1] It may be used in treating infections, inflammation, cold, fever and diarrhea.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Liu et al.
International immunopharmacology, 8(7), 951-958 (2008-05-20)
Andrographis paniculata Nees is an official herbal medicine for treatment of infection and inflammation in China. Andrograpanin, the one of diterpene lactones in A. paniculata, is a hydrolysate from neoandrographolide in vivo and in vitro. The goal of the present
Li-Li Ji et al.
Journal of cellular biochemistry, 95(5), 970-978 (2005-06-07)
Andrographis paniculata is a traditional Chinese medicine (TCM) that has been effectively used for treatment of infection, inflammation, cold, fever, and diarrhea in China. However, mechanism of its therapeutic function is not well known. In the current study, we showed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service