Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19367

Supelco

3-Deoxy-D-arabino-hexonic acid lithium salt

analytical standard

Synonym(s):

NSC 71039

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6 · xLi+
CAS Number:
Molecular Weight:
180.16 (free acid basis)
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

assay

≥98.0% (TLC)

form

solid

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

InChI

1S/C6H12O6/c7-2-5(10)3(8)1-4(9)6(11)12/h3-5,7-10H,1-2H2,(H,11,12)

InChI key

YGMNHEPVTNXLLS-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service