Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
HOCH2C≡CCH2OH
CAS Number:
Molecular Weight:
86.09
EC Number:
203-788-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1071237
MDL number:
vapor pressure
<0.1 mmHg ( 55 °C)
grade
purum
assay
≥98.0% (GC)
color
slightly brown
bp
238 °C (lit.)
mp
53-58 °C
SMILES string
OCC#CCO
InChI
1S/C4H6O2/c5-3-1-2-4-6/h5-6H,3-4H2
InChI key
DLDJFQGPPSQZKI-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under 2-Butyne-1,4-diol
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 2
flash_point_f
305.6 °F - closed cup
flash_point_c
152 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R A Jedrychowski et al.
Journal of applied toxicology : JAT, 12(2), 117-122 (1992-04-01)
2-Butyne-1,4-diol was given to male and female Wistar Imp:DAK rats by oral gavage for 28 consecutive days in daily doses of 1, 10 or 50 mg kg-1 day-1. After 28 days all animals were necropsied. Blood samples were obtained and
Małgorzata Kupczewska-Dobecka et al.
Medycyna pracy, 60(5), 347-357 (2009-12-17)
Derived No Effect Level (DNEL(inh)) has been set for occupational exposure to but-2-yno-1,4-diol according to REACH principles. Maximum allowable concentration (MAC) and DNEL(inh) have been compared. Experimental data from two inhalation studies on rats and three oral studies have been
Allergic contact dermatitis from 2-butin-1,4-diol.
V Blaschke et al.
Allergy, 56(3), 264-265 (2001-03-17)


