Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

18951

Supelco

1-Naphthyl isothiocyanate

derivatization grade (HPLC), LiChropur, ≥98.5% (HPLC and GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NCS
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (HPLC)

Quality Level

assay

≥98.5% (HPLC and GC)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

mp

55.5-57 °C (lit.)

SMILES string

S=C=Nc1cccc2ccccc12

InChI

1S/C11H7NS/c13-8-12-11-7-3-5-9-4-1-2-6-10(9)11/h1-7H

InChI key

JBDOSUUXMYMWQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhavana S Solanky et al.
Toxicological sciences : an official journal of the Society of Toxicology, 126(2), 306-316 (2012-01-21)
Certain human diseases affecting the biliary tree can be modeled in rats by ingestion of the hepatobiliary toxin alpha-naphthylisothiocyanate (ANIT). Phosphorus magnetic resonance spectroscopy (MRS) allows the noninvasive monitoring of cell dynamics through detection of phosphodiesters (PDE) and phosphomonoesters (PME).
Juhyun Park et al.
African journal of traditional, complementary, and alternative medicines : AJTCAM, 8(5 Suppl), 220-225 (2011-01-01)
This study evaluated the potential beneficial effect of Moutan Cortex Radicis (MCR) in a murine model of carbon tetrachloride (CCl(4))-, D-galactosamine (GalN)- and α-naphthylisothiocyanate (ANIT)-induced liver injury. Acute hepatotoxicity was induced by intraperitoneal injection of CCl(4) (10 µL/kg), GalN (700
Christina M Van Itallie et al.
Methods in molecular biology (Clifton, N.J.), 762, 1-11 (2011-07-01)
Tight junctions restrict the paracellular movement of ions, solutes, drugs, and larger material across epithelia and endothelia. For practical purposes, the barrier can be modeled as having two components. The first is a system of small 4  Å radius pores
Igor Sukhotnik et al.
Pediatric surgery international, 28(2), 161-169 (2011-10-13)
Progressive hyperbilirubinemia and end-stage liver failure are among the most serious complications of short bowel syndrome (SBS), representing the principle cause of death in a majority of fatal cases. In the current study, we examined the effects of alpha-naphthylisothiocyanate (ANIT)-induced
Fan Yang et al.
Journal of ethnopharmacology, 144(3), 599-604 (2012-10-13)
To evaluate the hepatoprotective effect of the root and rhizome of Rhubarb (Rhei Radix et Rhizoma) against α-naphthylisothiocyanate (ANIT)-induced liver injury using metabolic profile of bile acids (BAs) along with biochemical parameters and histological alterations. Ultra-performance liquid chromatography coupled with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service