18698
1-Methylpyrrole
analytical standard
Synonym(s):
N-Methylpyrrole
About This Item
grade
analytical standard
vapor density
2.8 (vs air)
vapor pressure
15 mmHg ( 20.2 °C)
assay
≥99.0% (GC)
shelf life
limited shelf life, expiry date on the label
expl. lim.
6 %
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤0.5% water
refractive index
n20/D 1.488-1.490
bp
112-113 °C (lit.)
mp
−57 °C (lit.)
density
0.914 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
Cn1cccc1
InChI
1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3
InChI key
OXHNLMTVIGZXSG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
60.8 °F - closed cup
flash_point_c
16 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service