Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

184527

Sigma-Aldrich

Pyridine

spectrophotometric grade, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein/REAXYS Number:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39151701
PubChem Substance ID:

grade

spectrophotometric grade

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

assay

≥99%

autoignition temp.

899 °F

expl. lim.

12.4 %

impurities

<0.100% water

evapn. residue

<0.002%

color

colorless

refractive index

n20/D 1.509 (lit.)

bp

115 °C (lit.)

mp

−42 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 315 nm Amax: 1.00
λ: 330 nm Amax: 0.15
λ: 335 nm Amax: 0.05
λ: 350-400 nm Amax: 0.01

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

68.0 °F - closed cup

flash_point_c

20 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicotine Levels and Presence of Selected Tobacco-Derived Toxins in Tobacco Flavoured Electronic Cigarette Refill Liquids.
Farsalinos KE, et al.
International Journal of Environmental Research and Public Health, 12(4), 3439-3452 (2015)
Daniel A Kirschner et al.
Current Alzheimer research, 5(3), 288-307 (2008-06-10)
Targeting the initial formation of amyloid assemblies is a preferred approach to therapeutic intervention in amyloidoses, which include such diseases as Alzheimer's, Parkinson's, Huntington's, etc., as the early-stage, oligomers that form before the development of beta-conformation-rich fibers are thought to
The far-infrared spectra of metal-halide complexes of pyridine and related ligands.
Clark RJH and Williams CS.
Inorganic Chemistry, 4(3), 350-357 (1965)
Total syntheses of complanadines A and B.
Le Zhao et al.
Angewandte Chemie (International ed. in English), 52(6), 1722-1725 (2013-01-03)
Nathalie Ségaud et al.
Inorganic chemistry, 52(2), 691-700 (2013-01-11)
We report the synthesis, characterization, and solution chemistry of a series of new Fe(II) complexes based on the tetradentate ligand N-methyl-N,N'-bis(2-pyridyl-methyl)-1,2-diaminoethane or the pentadentate ones N,N',N'-tris(2-pyridyl-methyl)-1,2-diaminoethane and N,N',N'-tris(2-pyridyl-methyl)-1,3-diaminopropane, modified by propynyl or methoxyphenyltriazolyl groups on the amino functions. Six of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service