18443
Bisphenol F diglycidyl ether
mixture of diastereomers, analytical standard
Synonym(s):
BFDGE, Bis[4-(glycidyloxy)phenyl]methane
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥95.0% (GC)
quality
mixture of diastereomers
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
C1OC1COc2ccc(Cc3ccc(OCC4CO4)cc3)cc2
InChI
1S/C19H20O4/c1-5-16(20-10-18-12-22-18)6-2-14(1)9-15-3-7-17(8-4-15)21-11-19-13-23-19/h1-8,18-19H,9-13H2
InChI key
XUCHXOAWJMEFLF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Canned food[4][5] and beverages using liquid chromatography coupled to tandem mass spectrometry (LC–MS/MS)[4] and high-performance liquid chromatography with fluorescence detection (HPLC-FLD).[5]
- Wastewaters using gas chromatography coupled to mass spectrometry (GC-MS).[6]
related product
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service