Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

182508

Supelco

Poly(vinyl acetate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(O2CCH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

mol wt

average Mw 124,800 (1st standard)
average Mw 194,800 (2nd standard)
average Mw 237,000 (3rd standard)

Quality Level

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)C=C

InChI

1S/C4H6O2/c1-3-6-4(2)5/h3H,1H2,2H3

InChI key

XTXRWKRVRITETP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl acetate) [PVAc] is a biocompatible and biodegradable polymer as it has hydrolyzable groups in the side chain,also it is non-toxic and non-carcinogenic.Owing to its biologically friendly nature , it is used in various biomedical application such as artificial organ implant, contact lens,cardiovascular devices and cartilage skin. It is also used in wound dressing and various drug-delivery applications.[1] It has been accepted as reference standard for universal calibration in gel permeation chromatography.[2]

Application

PVAc was used to prepare blends with poly(L-lactic acid) (PLA), in order to study the morphology of blends and miscibility system.[3]

Components

Set contains 3 standards; 25g of each Mw

Kit Components Only

Product No.
Description

  • average Mw 124,800, average Mn 52,700 25 g

  • average Mw 237,000, average Mn 89,900 25 g

  • average Mw 194,800, average Mn 63,600 25 g

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Composite poly(vinyl alcohol)/poly(vinyl acetate) electrospun nanofibrous mats as a novel wound dressing matrix for controlled release of drugs.
Jannesari M
International journal of nanomedicine, 6, 993-1003 (2011)
Poly (vinylacetate): reference materials and universal calibration in gel permeation chromatography.
Atkinson, C. M. L., and R. Dietz.
Eur. Polymer J., 15.1, 21-26 (1979)
Miscibility and morphology in blends of poly (L-lactic acid) and poly (vinyl acetate-co-vinyl alcohol).
Park, Jun Wuk, and Seung Soon Im.
Polymer, 44.15, 4341-4354 (2003)
Ehab A Fouad et al.
Expert opinion on drug delivery, 8(5), 693-703 (2011-03-23)
Developments in industrial pharmacy are often linked to the discovery of pharmaceutical excipients. Although recently introduced as a material for immediate release coatings, Kollicoat IR already has other applications. In this review, the different properties and pharmaceutical applications of Kollicoat
Susan R Sandeman et al.
Journal of colloid and interface science, 358(2), 582-592 (2011-04-05)
The textural and adsorption characteristics of a series of activated carbons (ACs), porous poly(vinyl alcohol) (PVA) gels, and PVA/AC composites were studied using scanning electron microscopy, mercury porosimetry, adsorption of nitrogen (at 77.4 K), cationic methylene blue (MB), anionic methyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service